Anti-S100 A4 Rabbit Polyclonal Antibody

51-026
10074-608EA 684.46 USD
10074-608
Anti-S100 A4 Rabbit Polyclonal Antibody
Antibodies
S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes. Suitable for use in Immunohistochemistry. Other applications have not been tested. Recognizes human S100A4.

S100A4 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, immunohistochemistry starting at 5 ?g/mL, immunofluorescence starting at 10 ?g/mL, and immunoprecipitation.

10 mM PBS, pH 7.6, 0.2% BSA, 15 mM sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: S100A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR