Anti-PD1 Rabbit Polyclonal Antibody

51-025
10074-606EA 491.23 USD
10074-606
Anti-PD1 Rabbit Polyclonal Antibody
PDCD1 antibody can be used in ELISA, and immunohistochemistry starting at 10 ?g/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.02% sodium azide.

Store PDCD1 antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type:
Antigen: PD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR