Anti-HSD11B2 Rabbit Polyclonal Antibody

51-002
10074-570EA 781.79 USD
10074-570
Anti-HSD11B2 Rabbit Polyclonal Antibody
Antibodies
HSD11B2 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

TBS

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HSD11B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gibbon, Gorilla, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR