Anti-SNCA Rabbit Polyclonal Antibody

Supplier: Prosci
50-241
10075-624EA 776.9 USD
10075-624
Anti-SNCA Rabbit Polyclonal Antibody
Antibodies
Alpha-synuclein is a presynaptic neuronal protein that is thought to be involved in the formation of SNARE complexes. Most significantly, aggregated alpha-synuclein is one of the major components found in the Lewy bodies that occur in Parkinson’s disease (PD) and other neurodegenerative disorders (Okochi et al., 2000). Mutation of the alpha-synuclein gene is associated with familial forms of PD (Newmann et al., 2002). Recent evidence suggests that phosphorylation of alpha-synuclein at ser129 plays a role in the formation of inclusions in PD (Smith et al., 2005; Masliah et al., 2005).

The antibody has been directly tested for reactivity in Western blots with rat tissue. It is anticipated that the antibody will react with bovine, canine, human, mouse, and non-human primate based on the fact that these species have 100% homology with the amino acid sequence used as antigen.

Affinity Purified

100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Snca
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR