Anti-SYN1 Rabbit Polyclonal Antibody

Supplier: Prosci

50-239
10075-698EA 582.92 USD
10075-698
Anti-SYN1 Rabbit Polyclonal Antibody
Antibodies
Synapsin I plays a key role in synaptic plasticity in brain (Feng et al., 2002; Nayak et al., 1996). This effect is due in large part to the ability of the synapsins to regulate the availability of synaptic vesicles for release. The role of synapsin in synaptic plasticity and in synaptogensis is regulated by phosphorylation (Jovanovic et al., 2001; Kao et al., 2002). Ser 549 along with Ser 62 and Ser 67 are the sites of synapsin I that are phosphorylated by MAP kinase (Jovanovic et al., 1996). Phosphorylation and subsequent dephosphorylation of this site is thought to play a key role in synaptic vesicle trafficking.

The antibody has been directly tested for reactivity in Western blots with rat tissue. It is anticipated that the antibody will react with human, mouse, non-human primate, bovine and canine tissues based on the fact that these species have 100% homology with the amino acid sequence used as antigen.

Affinity Purified

100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SYN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR