Anti-RPH3A Rabbit Polyclonal Antibody

Supplier: Prosci
50-236
10075-506EA 776.66 USD
10075-506
Anti-RPH3A Rabbit Polyclonal Antibody
Antibodies
Rabphilin-3A is a peripheral membrane protein that binds Ca2+ and phospholipids and is attached to synaptic vesicle membranes (Geppert et al., 1994;Li et al., 1994). The expression of rabphilin 3A is reduced in an animal model of Huntington’s disease(Smith et al., 2005). Rabphilin can be phosphorylated at Ser234 by both CAM kinase II and PKA and this phosphorylation has been suggested to regulate neuronal activity during development in a synapse-specific manner (Fykse et al., 1995;Foletti and Scheller, 2001).

The antibody has been directly tested for reactivity in Western blots with rat tissue. It is anticipated that the antibody will react with mouse based on the fact that this species has 100% homology with the amino acid sequence used as antigen.

Affinity Purified

100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Rph3a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR