Anti-GABBR1 Rabbit Polyclonal Antibody

Supplier: Prosci
50-227
10075-644EA 776.9 USD
10075-644
Anti-GABBR1 Rabbit Polyclonal Antibody
Antibodies
Gamma-aminobutyric acid (GABA) is the primary inhibitory neurotransmitter in the central nervous system. There are two major classes of GABA receptors: the GABAA and the GABAB subtype of receptors. GABAB receptors are heterodimeric G protein-coupled receptors that mediate slow synaptic inhibition in the central nervous system. Phosphorylation enhances GABAB receptor effector coupling (Couve et al., 2004). Phosphorylation of Ser923 is thought to be important in the regulation of GABAB receptor function.

The antibody has been directly tested for reactivity in Western blots with rat tissue. It is anticipated that the antibody will react with bovine, human, mouse and non-human primate tissues based on the fact that these species have 100% homology with the amino acid sequence used as antigen.

Affinity Purified

100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Gabbr1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR