Stargazin is a member of the transmembrane AMPAR regulatory proteins (TARP) family and is involved in glutamate receptor trafficking. It has been recently demonstrated (Tomita et al., 2005; Priel et al., 2005) that the interaction between stargazin and AMPA receptors is critical for the correct localization of the receptors at the synapse. Phosphorylation of the stargazin protein at Thr 321 by Protein Kinase A regulates its interaction with PSD-95 and synaptic targeting of AMPA receptors (Choi et al., 2002).
The antibody has been directly tested for reactivity in rat tissue. It is anticipated that the antibody will also work with mouse tissue based on the fact that this species has 100% homology with the amino acid sequence used as antigen.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: Cacng2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat