Anti-RXRB Mouse Monoclonal Antibody [clone: 147]

Supplier: Prosci
50-175
10075-650EA 776.9 USD
10075-650
Anti-RXRB Mouse Monoclonal Antibody [clone: 147]
Antibodies
Retinoic acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells (Joshi et al., 2005). The retinoid X receptors (RXRs) family (RXRalpha, beta and gamma), preferentially bind 9-cis-RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer (Abu et al., 2005). RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors (Lane and Bailey, 2005).

The antibody has been directly tested for reactivity in Western blots with mouse and rat tissue. It is anticipated that the antibody will react with human tissue based on the fact that this species has 100% homology with the amino acid sequence used as antigen.

Protein G purified

100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RXRB
Clonality: Monoclonal
Clone: 147
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR