Anti-Clvs1 Rabbit Polyclonal Antibody

Supplier: Prosci

50-153
10075-578EA 582.92 USD
10075-578
Anti-Clvs1 Rabbit Polyclonal Antibody
Antibodies
Affinity Purified

100 uL in PBS + 50% glycerol

Type: Primary
Antigen: Clvs1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR