Anti-CDK5 Mouse Polyclonal Antibody [clone: 1H3]

Supplier: Prosci
50-150
10075-582EA 776.9 USD
10075-582
Anti-CDK5 Mouse Polyclonal Antibody [clone: 1H3]
Antibodies
Protein G purified

100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.

Type: Primary
Antigen: Cdk5
Clonality: Monoclonal
Clone: 1H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR