Anti-Calb2 Sheep Polyclonal Antibody

Supplier: Prosci

50-149
10075-574EA 582.92 USD
10075-574
Anti-Calb2 Sheep Polyclonal Antibody
Antibodies
Protein A purified

100 uL in 10 mM Tris, 100 mM NaCl

Type: Primary
Antigen: Calb2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR