Anti-NR2C2 Rabbit Polyclonal Antibody

Supplier: Prosci
50-147
10075-576EA 701.93 USD
10075-576
Anti-NR2C2 Rabbit Polyclonal Antibody
Antibodies
Neat Serum

Type: Primary
Antigen: Nr2c2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR