Anti-TARDBP Rabbit Polyclonal Antibody

Supplier: Prosci
50-145
10075-710EA 776.9 USD
10075-710
Anti-TARDBP Rabbit Polyclonal Antibody
Antibodies
Affinity Purified

100 uL in PBS + 30% glycerol

Type: Primary
Antigen: TARDBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR