Anti-SYNJ1 Mouse Polyclonal Antibody [clone: 5H1]

Supplier: Prosci
50-143
10075-572EA 776.9 USD
10075-572
Anti-SYNJ1 Mouse Polyclonal Antibody [clone: 5H1]
Antibodies
Protein G purified

100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.

Type: Primary
Antigen: Synj1
Clonality: Monoclonal
Clone: 5H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR