Anti-POSTN Rabbit Polyclonal Antibody

Supplier: Prosci
50-138
10075-688EA 776.66 USD
10075-688
Anti-POSTN Rabbit Polyclonal Antibody
Antibodies
Affinity Purified

100 uL in PBS

Type: Primary
Antigen: Postn
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR