Anti-NUP98 Mouse Polyclonal Antibody [clone: 39C7]

Supplier: Prosci

50-132
10075-478EA 518.13 USD
10075-478
Anti-NUP98 Mouse Polyclonal Antibody [clone: 39C7]
Antibodies
Total IgG fraction

Type: Primary
Antigen: NUP98
Clonality: Monoclonal
Clone: 39C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR