Anti-Nefm Chicken Polyclonal Antibody

Supplier: Prosci
50-128
10075-686EA 701.93 USD
10075-686
Anti-Nefm Chicken Polyclonal Antibody
Antibodies
Total IgY fraction

Type: Primary
Antigen: Nefm
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR