Anti-AQP2 Rabbit Polyclonal Antibody

Supplier: Prosci
50-116
10075-560EA 776.9 USD
10075-560
Anti-AQP2 Rabbit Polyclonal Antibody
Antibodies
Affinity Purified

100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.

Type: Primary
Antigen: Aqp2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR