Anti-MARCKS Rabbit Polyclonal Antibody

50-108
10075-552EA 457.73 USD
10075-552
Anti-MARCKS Rabbit Polyclonal Antibody
Neat Serum

Type:
Antigen: MARCKS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR