Anti-COL1A1 Rabbit Polyclonal Antibody

Supplier: Prosci
50-106
10075-474EA 776.9 USD
10075-474
Anti-COL1A1 Rabbit Polyclonal Antibody
Antibodies
Affinity Purified

100 uL in PBS

Type: Primary
Antigen: COL1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR