Anti-Gstk1 Rabbit Polyclonal Antibody

Supplier: Prosci
50-102
10075-592EA 701.93 USD
10075-592
Anti-Gstk1 Rabbit Polyclonal Antibody
Antibodies
Serum

Type: Primary
Antigen: Gstk1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR