Anti-ERBB2 Rabbit Polyclonal Antibody

49-999
10074-568EA 674.53 USD
10074-568
Anti-ERBB2 Rabbit Polyclonal Antibody
Antibodies
ERBB2 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.

10 mM PBS, pH 7.4, 0.2% BSA, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ERBB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gibbon, Dog, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR