Anti-MUC13 Rabbit Polyclonal Antibody

Supplier: Prosci
49-990
10079-368EA 785.68 USD
10079-368
Anti-MUC13 Rabbit Polyclonal Antibody
Antibodies
Mucin 13 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MUC13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR