Anti-MFI2 Rabbit Polyclonal Antibody

49-987
10079-362EA 684.46 USD
10079-362
Anti-MFI2 Rabbit Polyclonal Antibody
Antibodies
P97 antibody can be used in Western Blot starting at 1 - 10 ?g/mL, immunohistochemistry starting at 10 ?g/mL, immunocytochemistry starting at 0.1 - 1 ?g/mL, and immunoprecipitation starting at 1 - 10 ?g/mL

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MFI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR