Anti-TFF2 Rabbit Polyclonal Antibody

49-985
10079-358EA 674.35 USD
10079-358
Anti-TFF2 Rabbit Polyclonal Antibody
Antibodies
Trefoil Factor 2 antibody can be used in ELISA, and immunohistochemistry starting at 10 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TFF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR