Anti-TH Rabbit Polyclonal Antibody

49-946
10079-326EA 684.46 USD
10079-326
Anti-TH Rabbit Polyclonal Antibody
Antibodies
Tyrosine hydroxylase in involved in the conversion of phenylalanine to dopamine. It is the rate-limiting enzyme in the synthesis of catecholamines and as such has a key role in the physiology of adrenergic neurons. TH is encoded by four distinct mRNAs produced by alternative splicing of a single primary transcript. Expression of the mRNAs varies in different parts of the nervous system. Defects in TH are the cause of autosomal recessive Segawa syndrome.

Tyrosine Hydroxylase antibody can be used in ELISA, Western Blot starting at 1:1000 - 1:3000, and immunohistochemistry starting at 10 ?g/mL.

50 uL of 10 mM HEPES, pH 7.5, 150 mM NaCl, 0.1 mg/mL BSA, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR