Anti-CK14 Mouse Monoclonal Antibody

49-931
10079-314EA 674.53 USD
10079-314
Anti-CK14 Mouse Monoclonal Antibody
Antibodies
KRT14 antibody can be used in ELISA, Western Blot starting at 1:1000 - 1:2000, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.4, 0.09% sodium azide, 0.2% BSA, before the addition of glycerol to 40%

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KRT14
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR