Anti-STAT5A Rabbit Polyclonal Antibody

Supplier: Prosci

49-925
10074-560EA 582.9 USD
10074-560
Anti-STAT5A Rabbit Polyclonal Antibody
Antibodies
STAT5 antibody can be used in ELISA, Western Blot starting at 2 - 4 ?g/mL, immunohistochemistry starting at 5 ?g/mL, and immunoprecipitation

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Stat5a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR