Beta 2-microglobulin is the low molecular weight component of HLA antigens and as such is involved in the presentation of endogenous antigens, activation of T lymphocytes and functional emergence of natural killer cells. It is found in the serum of normal individuals and in the urine in elevated amounts in patients with Wilson disease, cadmium poisoning and other conditions leading renal tubular dysfunction. ESTs have been isolated from libraries generated using virtually every tissue in the body.
B2M antibody can be used in immunohistochemistry starting at 20 ?g/mL.
DEAE-column Chromatography
PBS, pH 7.4, 15 mM sodium azide, before the addition of glycerol to 40%
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human