Anti-PLTP Rabbit Polyclonal Antibody

49-918
10076-576EA 698.16 USD
10076-576
Anti-PLTP Rabbit Polyclonal Antibody
Antibodies
PLTP antibody can be used in ELISA, Western Blot starting at 1:1000 - 1:2000, and immunohistochemistry starting at 5 ?g/mL.

PBS, 1 % BSA, 0.02% Thimerosal.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PLTP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR