Anti-TPM1 Mouse Monoclonal Antibody

49-879
10076-562EA 698.16 USD
10076-562
Anti-TPM1 Mouse Monoclonal Antibody
Antibodies
TPM1 antibody can be used in ELISA, Western Blot starting at 1:250 - 1:2000, and immunohistochemistry starting at 5 ?g/mL.

10 mM PBS, pH 7.4, 0.2% BSA, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TPM1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Chicken, Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR