Anti-NEDD4 Rabbit Polyclonal Antibody

49-851
10076-532EA 540.97 USD
10076-532
Anti-NEDD4 Rabbit Polyclonal Antibody
NEDD4 antibody can be used in ELISA, Western Blot starting at 1:1000 - 1:2000, and immunohistochemistry starting at 10 ?g/mL.

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide, 30% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NEDD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR