Anti-SLC3A2 Mouse Monoclonal Antibody [clone: [MEM-108]]

49-812
10076-518EA 674.53 USD
10076-518
Anti-SLC3A2 Mouse Monoclonal Antibody [clone: [MEM-108]]
Antibodies
CD98 antibody can be used in ELISA starting at 0.1 - 1 ?g/mL, Western Blot starting at 1 ?g/mL, immunohistochemistry starting at 5 ?g/mL, and immunohistochemistry in frozen tissues starting at 5 ?g/mL.

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SLC3A2
Clonality: Monoclonal
Clone: [MEM-108]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR