Anti-HP Rabbit Polyclonal Antibody

Supplier: Prosci

49-808
10076-512EA 582.9 USD
10076-512
Anti-HP Rabbit Polyclonal Antibody
Antibodies
Suitable for use in ELISA. Other applications not tested. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-rabbit serum.

PBS, pH 7.2, 0.01% sodium azide. No stabilizing proteins added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR