Anti-CK15 Mouse Monoclonal Antibody [clone: [15K01]]

49-804
10076-504EA 684.46 USD
10076-504
Anti-CK15 Mouse Monoclonal Antibody [clone: [15K01]]
Antibodies
KRT15 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 10 ?g/mL, and flow cytometry. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0 for 10-20 min., followed by cooling at RT for 20 min. Immunofluorescence, Western Blotting, Immunohistochemistry Recognizes keratin 15.

PBS, pH 7.4, 0.2% BSA, 15 mM sodium azide, before the addition of glycerol to 40%

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KRT15
Clonality: Monoclonal
Clone: [15K01]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR