Anti-MBP Mouse Monoclonal Antibody

Supplier: Prosci

49-792
10074-548EA 665.41 USD
10074-548
Anti-MBP Mouse Monoclonal Antibody
Antibodies
Suitable for use in ELISA, Western Blot, Immunohistochemistry and quantification of myelin basic protein. Other applications have not been tested. Recognizes residues 130-136 in human myelin basic protein (21.5kD and 18.5kD molecular forms). Species Crossreactivity: Primate, rabbit, sheep, goat, rat and mouse. The classic group of MBP isoforms (isoform 4-isoform 14) are with PLP the most abundant protein components of the myelin membrane in the CNS. They have a role in both its formation and stabilization. The smaller isoforms might have an important role in remyelination of denuded axons in multiple sclerosis. The non-classic group of MBP isoforms (isoform 1-isoform 3/Golli-MBPs) found in bone marrow and the immune system may preferentially have a role in the early developing brain long before myelination, maybe as components of transcriptional complexes, and may also be involved in signaling pathways in T-cells and neural cells.

  • Myelin Basic Protein antibody can be used in immunocytochemistry starting at 10 μg/mL, immunohistochemistry starting at 10 μg/mL, and flow cytometry.

Protein G Column

PBS, pH 7.4, contains no preservative.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MBP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Goat, Human, Mouse, Primate, Rabbit, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR