Anti-SAA1 Mouse Monoclonal Antibody

49-782
10074-536EA 674.35 USD
10074-536
Anti-SAA1 Mouse Monoclonal Antibody
Antibodies
SAA1 antibody can be used in immunohistochemistry starting at 20 ?g/mL.

PBS, pH 7.2.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SAA1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR