Anti-CA2 Rabbit Polyclonal Antibody

49-763
10074-508EA 1000.72 USD
10074-508
Anti-CA2 Rabbit Polyclonal Antibody
Antibodies
Pe HRPCA2 is one of several (at least 7) isozymes of carbonic anhydrase. Carbonic anhydrase catalyzes reversible hydration of carbon dioxide. Defects in this enzyme are associated with osteopetrosis and renal tubular acidosis.

Carbonic Anhydrase 2 antibody can be used in ELISA starting at 1:5000 - 1:10000, Western Blot, immunohistochemistry starting at 2.5 ?g/mL, and immunoprecipitation

Ion Exchange Chromatography

PBS, pH 7.2, 0.01% Gentamicin sulfate, 10 mg/mL BSA, before the addition of glycerol to 40%

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR