Anti-KAL1 Rabbit Polyclonal Antibody

49-752
10079-296EA 674.53 USD
10079-296
Anti-KAL1 Rabbit Polyclonal Antibody
Antibodies
Mutations in the KAL1 gene cause the X-linked Kallmann syndrome. The predictated KAL1 protein sequence is similar to proteins known to function in neural cell adhesion and axonal migration.

KAL1 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 1:500, immunofluorescence, and immunoprecipitation.

Protein G Column

PBS, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KAL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR