Anti-THPO Rabbit Polyclonal Antibody

Supplier: Prosci

49-744
10079-280EA 582.9 USD
10079-280
Anti-THPO Rabbit Polyclonal Antibody
Antibodies
Thrombopoietin antibody can be used in immunohistochemistry starting at 5 ?g/mL, immunoprecipitation, and flow cytometry.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: THPO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR