Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAGEA9 Rabbit Polyclonal Antibody

Supplier: Prosci

49-716
10079-226EA 515.21 USD InStock
10079-226
Anti-MAGEA9 Rabbit Polyclonal Antibody
Antibodies
MAGEA9 antibody can be used in ELISA starting at 1:20000 - 1:100000, Western Blot starting at 1:2000 - 1:8000, immunohistochemistry starting at 5 - 10 μg/mL, and immunofluorescence.

Protein G Column

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MAGEA9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty