Anti-MAGEA1 Rabbit Polyclonal Antibody

49-710
10076-486EA 540.97 USD
10076-486
Anti-MAGEA1 Rabbit Polyclonal Antibody
MAGE was originally isolated from human melanoma cells as an antigen that is recognizes by cytolytic T lymphocytes. It has since been documented in other carcinomas. In addition, expression has been documented in highly proliferating germ cells.

MAGEA1 antibody can be used in ELISA starting at 1:10000 - 1:40000, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 2.5 ?g/mL.

Protein G Column

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MAGEA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR