Anti-MLL Rabbit Polyclonal Antibody

49-695
10079-216EA 540.97 USD
10079-216
Anti-MLL Rabbit Polyclonal Antibody
MLL antibody can be used in ELISA starting at 1:4000 - 1:20000, Western Blot starting at 1:500 - 1:3000, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MLL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR