Anti-PRKACB Rabbit Polyclonal Antibody

Supplier: Prosci

49-689
10079-204EA 582.9 USD
10079-204
Anti-PRKACB Rabbit Polyclonal Antibody
Antibodies
PRKACGB, a cAMP type protein kinase, is the catalytic beta subunit for the PKA holoenzyme. Multiple isoforms of PRKACB have been reported.

Protein Kinase A antibody can be used in ELISA starting at 1:1000 - 1:5000, Western Blot starting at 1:250 - 1:2000, and immunohistochemistry starting at 5 ?g/mL.

Protein G Column

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PRKACB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR