Anti-MMP27 Rabbit Polyclonal Antibody

Supplier: ProSci
49-682
10079-190EA 674.53 USD
10079-190
Anti-MMP27 Rabbit Polyclonal Antibody
Antibodies
MMP27 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 ?g/mL, immunofluorescence, and immunoprecipitation.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MMP27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR