Anti-GDF3 Rabbit Polyclonal Antibody

Supplier: ProSci
49-671
10079-170EA 674.53 USD
10079-170
Anti-GDF3 Rabbit Polyclonal Antibody
Antibodies
GDF3 antibody can be used in ELISA starting at 1:10000 - 1:50000, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GDF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR