Anti-GDF15 Rabbit Polyclonal Antibody

49-669
10079-166EA 540.97 USD
10079-166
Anti-GDF15 Rabbit Polyclonal Antibody
GDF15 antibody can be used in ELISA starting at 1:2000 - 1:10000, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 2.5 ?g/mL, and immunofluorescence.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GDF15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR