Anti-AFF1 Rabbit Polyclonal Antibody

Supplier: Prosci

49-658
10079-148EA 582.9 USD
10079-148
Anti-AFF1 Rabbit Polyclonal Antibody
Antibodies
AFF1 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 1.25 ?g/mL, and immunoprecipitation

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: AFF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR