Anti-BMP6 Rabbit Polyclonal Antibody

49-650
10079-132EA 540.97 USD
10079-132
Anti-BMP6 Rabbit Polyclonal Antibody
Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. Through molecular cloning, 7 BMP cDNAs, designated BMP1 through BMP7, have been recovered. BMP2 through BMP7, are members of the transforming growth factor-beta superfamily of regulatory molecules. From a high degree of amino acid sequence homology, BMP5, BMP6, and BMP7 are recognized as a subfamily of the BMPs.

BMP6 antibody can be used in ELISA starting at 1:10000 - 1:50000, Western Blot starting at 1:500 - 1:3000, immunohistochemistry starting at 2.5 ?g/mL, and immunofluorescence.

Protein G Column

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BMP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR